N-Chloroacetyl-3-aminophenylpropionic acid ≥95%

Supplier: Matrix Scientific

054696-500MG 054696-1G
101908-170EA 556.92 USD
101908-170 101908-172
N-Chloroacetyl-3-aminophenylpropionic acid ≥95%
N-Chloroacetyl-3-aminophenylpropionic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
MDL Number: MFCD16140271
CAS Number: 1221792-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR