N-Chloroacetyl-3-aminophenylacetic acid ≥95%

Supplier: Matrix Scientific

054695-500MG 054695-1G
101908-166EA 529.23 USD
101908-166 101908-168
N-Chloroacetyl-3-aminophenylacetic acid ≥95%
N-Chloroacetyl-3-aminophenylacetic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₃
MDL Number: MFCD16140270
CAS Number: 1221792-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR