{4-[(Chloroacetyl)amino]phenyl}acetic acid ≥95%

Supplier: Matrix Scientific

054694-500MG 054694-1G
101908-162EA 529.23 USD
101908-162 101908-164
{4-[(Chloroacetyl)amino]phenyl}acetic acid ≥95%
{4-[(Chloroacetyl)amino]phenyl}acetic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₃
MDL Number: MFCD08753651
CAS Number: 90798-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR