Boc-DL-3,3-diphenylalanine ≥95%

Supplier: Matrix Scientific

054687-500MG 054687-1G
101908-132EA 556.92 USD
101908-132 101908-134
Boc-DL-3,3-diphenylalanine ≥95%
Boc-DL-3,3-diphenylalanine

Hazard: Irritant

Formula: C₂₀H₂₃NO₄
MW: 341.41 g/mol
MDL Number: MFCD00797562
CAS Number: 119363-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR