3-(1H-Benzimidazol-1-yl)propylamine ≥95%

Supplier: Matrix Scientific

054680-500MG 054680-1G
101908-102EA 375.38 USD
101908-102 101908-104
3-(1H-Benzimidazol-1-yl)propylamine ≥95%
3-(1H-Benzimidazol-1-yl)propylamine

Hazard: Irritant

Formula: C₁₀H₁₃N₃
MDL Number: MFCD03093044
CAS Number: 73866-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR