3-Benzenesulfonylpropylamine hydrochloride ≥95%

Supplier: Matrix Scientific

054679-500MG 054679-1G
101908-098EA 556.92 USD
101908-098 101908-100
3-Benzenesulfonylpropylamine hydrochloride ≥95%
3-Benzenesulfonylpropylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO₂S
MW: 235.73 g/mol
MDL Number: MFCD03840164
CAS Number: 98510-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR