Boc(aminophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-((tert-Butoxycarbonyl)amino)phenyl)acetic acid

054675-500MG 054675-1G
101908-084EA 58.46 USD
101908-084 101908-086
Boc(aminophenyl)acetic acid ≥95%
Boc(aminophenyl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MDL Number: MFCD09750482
CAS Number: 135807-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR