3-Aminoethyl-1-methylpyrazole dihydrochloride ≥95%

Supplier: Matrix Scientific

054670-500MG 054670-1G
101908-066EA 538.46 USD
101908-066 101908-068
3-Aminoethyl-1-methylpyrazole dihydrochloride ≥95%
3-Aminoethyl-1-methylpyrazole dihydrochloride

Hazard: Irritant

Formula: C₆H₁₃Cl₂N₃
MDL Number: MFCD16090012
CAS Number: 1221792-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR