4-Amino-N-[2-(dimethylamino)ethyl]benzamide ≥95%

Supplier: Matrix Scientific

054668-500MG 054668-1G
101908-058EA 556.92 USD
101908-058 101908-060
4-Amino-N-[2-(dimethylamino)ethyl]benzamide ≥95%
4-Amino-N-[2-(dimethylamino)ethyl]benzamide

Hazard: Irritant

Formula: C₁₁H₁₇N₃O
MW: 207.28 g/mol
MDL Number: MFCD01666732
CAS Number: 188758-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR