4-[4-(Acetylamino)phenoxy]butanoic acid ≥95%

Supplier: Matrix Scientific

054662-500MG 054662-1G
101908-034EA 529.23 USD
101908-034 101908-036
4-[4-(Acetylamino)phenoxy]butanoic acid ≥95%
4-[4-(Acetylamino)phenoxy]butanoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD08696414
CAS Number: 937600-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR