4-(2-Acetamidophenoxy)butanoic acid ≥95%

Supplier: Matrix Scientific

054660-500MG 054660-1G
101908-026EA 375.38 USD
101908-026 101908-028
4-(2-Acetamidophenoxy)butanoic acid ≥95%
4-(2-Acetamidophenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD09816326
CAS Number: 1016689-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR