(4-Acetamidophenoxy)acetic acid ≥95%

Supplier: Matrix Scientific

054658-500MG 054658-1G
101908-018EA 375.38 USD
101908-018 101908-020
(4-Acetamidophenoxy)acetic acid ≥95%
(4-Acetamidophenoxy)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD01242445
CAS Number: 39149-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR