4-(2-Acetamidoethyl)phenoxyacetic acid ≥95%

Supplier: Matrix Scientific

054657-500MG 054657-1G
101908-014EA 556.92 USD
101908-014 101908-016
4-(2-Acetamidoethyl)phenoxyacetic acid ≥95%
4-(2-Acetamidoethyl)phenoxyacetic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD16090032
CAS Number: 1221574-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR