5-Bromo-2-methylbenzotrifluoride ≥98%

Supplier: Matrix Scientific

054612-5G 054612-25G
101907-922EA 58.46 USD
101907-922 101907-924
5-Bromo-2-methylbenzotrifluoride ≥98%
5-Bromo-2-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631587
CAS Number: 86845-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR