4-Methyl-3-(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

054607-5G 054607-25G
101907-912EA 29.23 USD
101907-912 101907-914
4-Methyl-3-(trifluoromethyl)benzoic acid ≥98%
4-Methyl-3-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 175-178°C
MDL Number: MFCD01631592
CAS Number: 261952-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR