2-Bromo-3-fluorophenylacetic acid ≥98%

Supplier: Matrix Scientific

054605-1G 054605-5G
101907-904EA 95.38 USD
101907-904 101907-906
2-Bromo-3-fluorophenylacetic acid ≥98%
2-Bromo-3-fluorophenylacetic acid

Hazard: Irritant

Formula: C₈H₆BrFO₂
MDL Number: MFCD09864672
CAS Number: 958454-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR