5-Chloro-2-hydroxynicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-hydroxy-3-pyridinecarboxylic Acid

054604-5G 054604-25G
101907-900EA 64.62 USD
101907-900 101907-902
5-Chloro-2-hydroxynicotinic acid ≥98%
5-Chloro-2-hydroxynicotinic acid

Hazard: Irritant

Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
Melting Pt: 258…260 °C
MDL Number: MFCD00204158
CAS Number: 38076-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR