5-Bromo-2-oxo-1,2-dihydropyridine-3-carboxylic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-hydroxynicotinic acid, 5-Bromo-2-hydroxy-3-pyridinecarboxylic acid
054603-5G
054603-25G
054603-100G
101907-894EA
26.15
USD
101907-894
101907-896
101907-898
5-Bromo-2-oxo-1,2-dihydropyridine-3-carboxylic acid ≥98%
5-Bromo-2-oxo-1,2-dihydropyridine-3-carboxylic acid
Hazard: Irritant/Refrigerate
Formula:
C₆H₄BrNO₃ MW: 218.49 g/mol Boiling Pt: 354°C Melting Pt: 245 °C Flash Pt: 168°C |
MDL Number:
MFCD07363801 CAS Number: 104612-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...