5-Hydroxynorbornane-2,6-lactone ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one

054600-5G 054600-25G
101907-886EA 92.31 USD
101907-886 101907-888
5-Hydroxynorbornane-2,6-lactone ≥98%
5-Hydroxynorbornane-2,6-lactone

Hazard: Irritant

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD00988184
CAS Number: 92343-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR