1-Benzyl-3-methylpiperazine ≥97%

Supplier: Matrix Scientific

054591-1G 054591-5G
101907-874EA 150.77 USD
101907-874 101907-876
1-Benzyl-3-methylpiperazine ≥97%
1-Benzyl-3-methylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD01570796
CAS Number: 3138-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR