5-Bromo-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-bromo-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridine
054579-500MG
054579-1G
101907-840EA
753.85
USD
101907-840
101907-842
5-Bromo-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
5-Bromo-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine
Hazard: Irritant
Formula:
C₉H₉BrN₄ MW: 253.11 g/mol |
MDL Number:
MFCD14584777 CAS Number: 1221792-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...