5-Bromo-2-chloro-6-cyclopropylnicotinonitrile ≥95%

Supplier: Matrix Scientific

054578-500MG 054578-1G
101907-836EA 747.69 USD
101907-836 101907-838
5-Bromo-2-chloro-6-cyclopropylnicotinonitrile ≥95%
5-Bromo-2-chloro-6-cyclopropylnicotinonitrile

Hazard: Irritant

Formula: C₉H₆BrClN₂
MW: 257.52 g/mol
MDL Number: MFCD14584776
CAS Number: 1221792-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR