2-Chloro-1-iodo-5-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-iodo-4-nitrobenzene, 4-Chloro-3-iodonitrobenzene

054512-500MG 054512-1G
101907-576EA 186.15 USD
101907-576 101907-578
2-Chloro-1-iodo-5-nitrobenzene ≥95%
2-Chloro-1-iodo-5-nitrobenzene

Hazard: Irritant/Refrigerate

Formula: C₆H₃ClINO₂
MW: 283.4500 g/mol
MDL Number: MFCD11521301
CAS Number: 74534-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR