2-Chloro-4-(trifluoromethyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

054511-500MG 054511-1G
101907-572EA 600 USD
101907-572 101907-574
2-Chloro-4-(trifluoromethyl)nicotinonitrile ≥95%
2-Chloro-4-(trifluoromethyl)nicotinonitrile

Hazard: Irritant

Formula: C₇H₂ClF₃N₂
MDL Number: MFCD14584807
CAS Number: 896447-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR