Methyl-2,5,6-trichloroisonicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2,3,5-trichloroisonicotinate
054508-500MG
054508-1G
054508-5G
101907-560EA
556.92
USD
101907-560
101907-562
101907-564
Methyl-2,5,6-trichloroisonicotinate ≥95%
Methyl 2,5,6-trichloroisonicotinate
Hazard: Irritant
Formula:
C₇H₄Cl₃NO₂ MW: 240.4712 g/mol |
MDL Number:
MFCD14584804 CAS Number: 1221791-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...