cis-3,4-Diethylpyrrolidine-3,4-dicarboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

054507-500MG 054507-1G
101907-556EA 921.54 USD
101907-556 101907-558
cis-3,4-Diethylpyrrolidine-3,4-dicarboxylate hydrochloride ≥95%
cis-3,4-Diethylpyrrolidine-3,4-dicarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₄
Melting Pt: >112 °C (dec.)
MDL Number: MFCD14584803
CAS Number: 1235297-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR