2-Bromo-3,5,6-trifluoropyridine ≥98%

Supplier: Matrix Scientific

054393-1G 054393-5G
101907-250EA 2172.31 USD
101907-250 101907-252
2-Bromo-3,5,6-trifluoropyridine ≥98%
2-Bromo-3,5,6-trifluoropyridine

Hazard: Irritant

Formula: C₅HBrF₃N
MDL Number: MFCD12031583
CAS Number: 1186194-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR