2-Chloro-3-iodo-4-methylpyridine ≥98%

Supplier: Matrix Scientific

054390-1G 054390-5G 054390-25G
101907-240EA 49.23 USD
101907-240 101907-242 101907-244
2-Chloro-3-iodo-4-methylpyridine ≥98%
2-Chloro-3-iodo-4-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClIN
MDL Number: MFCD11878566
CAS Number: 926922-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR