5'-Bromo-2'-fluoroacetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(5-Bromo-2-fluorophenyl)ethanone
054340-1G
054340-5G
054340-25G
101907-124EA
21.54
USD
101907-124
102551-412
102551-414
5'-Bromo-2'-fluoroacetophenone ≥95%
5'-Bromo-2'-fluoroacetophenone
Hazard: Irritant
Formula:
C₈H₆BrFO MW: 217.04 g/mol |
MDL Number:
MFCD11226880 CAS Number: 198477-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...