2-(1H-1,2,4-Triazol-1-yl)benzoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

054338-500MG 054338-1G
101907-114EA 538.46 USD
101907-114 101907-116
2-(1H-1,2,4-Triazol-1-yl)benzoic acid hydrochloride ≥95%
2-(1H-1,2,4-Triazol-1-yl)benzoic acid hydrochloride

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
MDL Number: MFCD14581678
CAS Number: 1203898-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR