7H-Pyrrolo[2,3-d]pyrimidin-4-aminesulfuric acid ≥95%

Supplier: Matrix Scientific

054335-500MG 054335-1G
101907-102EA 3450.77 USD
101907-102 101907-104
7H-Pyrrolo[2,3-d]pyrimidin-4-aminesulfuric acid ≥95%
7H-Pyrrolo[2,3-d]pyrimidin-4-aminesulfuric acid

Hazard: Irritant

Formula: C₆H₈N₄O₄S
MW: 232.22 g/mol
MDL Number: MFCD09787504
CAS Number: 856600-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR