(S)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid ≥90%

Supplier: Matrix Scientific

Synonyms: Cbz-Tle-OH, Z-Tle-OH

054333-1G 054333-5G
101907-096EA 89.23 USD
101907-096 101907-098
(S)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid ≥90%
(S)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MW: 265.3 g/mol
MDL Number: MFCD00137411
CAS Number: 62965-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR