(Z)-3-Bromo-4-methoxy-4-oxo-2-butenoic acid ≥95%

Supplier: Matrix Scientific

054328-500MG 054328-1G
101907-078EA 278.46 USD
101907-078 101907-080
(Z)-3-Bromo-4-methoxy-4-oxo-2-butenoic acid ≥95%
(Z)-3-Bromo-4-methoxy-4-oxo-2-butenoic acid

Hazard: Irritant

Formula: C₅H₅BrO₄
MDL Number: MFCD14560536
CAS Number: 122457-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR