Ethyl-4-amino-2-chlorobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-amino-2-chlorobenzoate
054322-500MG
054322-1G
101907-064EA
181.54
USD
101907-064
101907-066
Ethyl-4-amino-2-chlorobenzoate ≥95%
Ethyl 4-amino-2-chlorobenzoate
Hazard: Irritant
Formula:
C₉H₁₀ClNO₂ MW: 199.6342 g/mol |
MDL Number:
MFCD09034640 CAS Number: 16017-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...