2-Fluoro-6-(methylamino)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-6-(methylamino)benzonitrile

054317-500MG 054317-1G 054317-5G
101907-042EA 113.85 USD
101907-042 101907-044 101907-046
2-Fluoro-6-(methylamino)benzenecarbonitrile ≥95%
2-Fluoro-6-(methylamino)benzenecarbonitrile

Hazard: Irritant

Formula: C₈H₇FN₂
MW: 150.16 g/mol
MDL Number: MFCD12193344
CAS Number: 96783-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR