5-Iodo-6-isopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

054313-500MG 054313-1G
101907-024EA 375.38 USD
101907-024 101907-026
5-Iodo-6-isopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
5-Iodo-6-isopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₉H₉IN₂O
MDL Number: MFCD14581669
CAS Number: 1203898-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR