N-Methyl-6-nitro-1,3-benzoxazol-2-amine ≥95%

Supplier: Matrix Scientific

054312-500MG 054312-1G
101907-020EA 375.38 USD
101907-020 101907-022
N-Methyl-6-nitro-1,3-benzoxazol-2-amine ≥95%
N-Methyl-6-nitro-1,3-benzoxazol-2-amine

Hazard: Irritant

Formula: C₈H₇N₃O₃
MDL Number: MFCD14581668
CAS Number: 78749-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR