2-Chloro-N'-hydroxy-3-pyridinecarboximidamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-N'-hydroxynicotinimidamide

054306-500MG 054306-1G 054306-5G
101907-000EA 278.46 USD
101907-000 101907-002 101907-004
2-Chloro-N'-hydroxy-3-pyridinecarboximidamide ≥95%
2-Chloro-N'-hydroxy-3-pyridinecarboximidamide

Hazard: Irritant

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD08273500
CAS Number: 468068-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR