Methyl-3-amino-2-(benzyloxy)-5-methylbenzoate hydrochloride ≥95%

Supplier: Matrix Scientific

054304-500MG 054304-1G
101906-994EA 556.92 USD
101906-994 101906-996
Methyl-3-amino-2-(benzyloxy)-5-methylbenzoate hydrochloride ≥95%
Methyl-3-amino-2-(benzyloxy)-5-methylbenzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈ClNO₃
MW: 307.78 g/mol
MDL Number: MFCD14560548
CAS Number: 1203898-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR