Methyl-3-amino-2-methoxy-5-methylbenzoate hydrochloride ≥95%
Supplier: Matrix Scientific
054303-500MG
054303-1G
101906-990EA
747.69
USD
101906-990
101906-992
Methyl-3-amino-2-methoxy-5-methylbenzoate hydrochloride ≥95%
Methyl-3-amino-2-methoxy-5-methylbenzoate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₄ClNO₃ |
MDL Number:
MFCD14560551 CAS Number: 1203898-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...