2-Bromo-4-nitro-6-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-bromo-5-nitrobenzotrifluoride

054302-500MG 054302-1G 054302-5G
101906-984EA 278.46 USD
101906-984 101906-986 101906-988
2-Bromo-4-nitro-6-(trifluoromethyl)aniline ≥95%
2-Bromo-4-nitro-6-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄BrF₃N₂O₂
MW: 285.0200 g/mol
MDL Number: MFCD14560537
CAS Number: 400-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR