2-Iodo-4-nitro-6-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-iodo-5-nitrobenzotrifluoride

054298-500MG 054298-1G 054298-5G
101906-964EA 375.38 USD
101906-964 101906-966 101906-968
2-Iodo-4-nitro-6-(trifluoromethyl)aniline ≥95%
2-Iodo-4-nitro-6-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄F₃IN₂O₂
MDL Number: MFCD14560547
CAS Number: 400-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR