Methyl-2-hydroxy-5-methyl-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

054296-500MG 054296-1G 054296-5G
101906-956EA 375.38 USD
101906-956 101906-958 101906-960
Methyl-2-hydroxy-5-methyl-3-nitrobenzenecarboxylate ≥95%
Methyl-2-hydroxy-5-methyl-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₉H₉NO₅
MW: 211.18 g/mol
MDL Number: MFCD14560553
CAS Number: 67191-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR