6-Acetylpicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Acetylpyridine-2-carboxylic acid

054294-500MG 054294-1G
101906-950EA 515.38 USD
101906-950 101906-952
6-Acetylpicolinic acid ≥95%
6-Acetylpicolinic acid

Hazard: Irritant

Formula: C₈H₇NO₃
MDL Number: MFCD09800364
CAS Number: 122637-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR