3-Chloro-4-(trifluoromethyl)-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054293-250MG 054293-1G
101906-946EA 470.77 USD
101906-946 101906-948
3-Chloro-4-(trifluoromethyl)-2-pyridine carboxylic acid ≥95%
3-Chloro-4-(trifluoromethyl)-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
MDL Number: MFCD14581664
CAS Number: 796090-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR