Ethyl-6-acetyl-2-pyridinecarboxylate ≥97%

Supplier: Matrix Scientific

054286-500MG 054286-1G 054286-5G
101906-916EA 323.08 USD
101906-916 101906-918 101906-920
Ethyl-6-acetyl-2-pyridinecarboxylate ≥97%
Ethyl-6-acetyl-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
Melting Pt: 49…51 °C
MDL Number: MFCD09261291
CAS Number: 114578-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR