Methyl-4-(formylamino)-3-methoxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

054284-500MG 054284-1G
101906-906EA 747.69 USD
101906-906 101906-908
Methyl-4-(formylamino)-3-methoxybenzenecarboxylate ≥95%
Methyl-4-(formylamino)-3-methoxybenzenecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD09991719
CAS Number: 700834-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR