6-Chloro-7-nitro-2H-1,4-benzoxazin-3(4H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Chloro-7-nitro-2H-benzo[b][1,4]oxazin-3(4H)-one
054281-500MG
054281-1G
054281-5G
101906-894EA
278.46
USD
101906-894
101906-896
101906-898
6-Chloro-7-nitro-2H-1,4-benzoxazin-3(4H)-one ≥95%
6-Chloro-7-nitro-2H-1,4-benzoxazin-3(4H)-one
Hazard: Irritant
Formula:
C₈H₅ClN₂O₄ MW: 228.59 g/mol |
MDL Number:
MFCD02660643 CAS Number: 116862-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...