Ethyl-5-chloro-3-(trifluoromethyl)-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

054278-500MG 054278-1G 054278-5G
101906-876EA 792.31 USD
101906-876 101906-878 101906-880
Ethyl-5-chloro-3-(trifluoromethyl)-2-pyridinecarboxylate ≥95%
Ethyl-5-chloro-3-(trifluoromethyl)-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₇ClF₃NO₂
MW: 253.61 g/mol
MDL Number: MFCD14581656
CAS Number: 1198475-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR