(4-Methyl-1,3-thiazol-5-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-5-thiazolemethanol, (4-Methylthiazol-5-yl)methanol

054276-500MG 054276-1G
101906-868EA 375.38 USD
101906-868 101906-870
(4-Methyl-1,3-thiazol-5-yl)methanol ≥95%
(4-Methyl-1,3-thiazol-5-yl)methanol

Hazard: Irritant

Formula: C₅H₇NOS
MW: 129.18 g/mol
Boiling Pt: 113-114°C
Melting Pt: 65…66 °C
MDL Number: MFCD09032911
CAS Number: 1977-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR